Learn More
Meso Scale Discovery UPLEX NHP TGFB2 ASY384well
SDP
Catalog No. 5000080269
Encompass
The Meso Scale Discovery U-PLEX NHP TGF-beta2 384-well assay kit is a multiplex immunoassay for measuring active and total TGF-beta2 in human serum plasma and cell culture supernatants It uses electrochemiluminescence detection for high sensitivity broad dynamic range and rapid results The kit includes a 384-well plate with capture and detection antibodies calibrators controls diluent read buffer and wash buffer Sensitivity is 0 3 pg/mL with an upper quantification limit of 10 000 pg/mL enabling high-throughput screening with a 12 5 L sample volume per well
Catalog No. 50-000-80269 Supplier Meso Scale Discovery Supplier No. K256XUK4
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX NHP TGF-beta2 384-well assay kit is a multiplex immunoassay for measuring active and total TGF-beta2 in human serum plasma and cell culture supernatants It uses electrochemiluminescence detection for high sensitivity broad dynamic range and rapid results The kit includes a 384-well plate with capture and detection antibodies calibrators controls diluent read buffer and wash buffer Sensitivity is 0 3 pg/mL with an upper quantification limit of 10 000 pg/mL enabling high-throughput screening with a 12 5 L sample volume per well