Learn More
Meso Scale Discovery UPLEX NHP TGFB2 ASYQP 25 PL
SDP
Catalog No. 5000080742
Encompass
The Meso Scale Discovery U-PLEX NHP TGF-beta 2 Assay Kit is a multiplex immunoassay that measures active and total TGF-beta 2 levels in non-human primate serum plasma and cell culture supernatants This electrochemiluminescent assay utilizes MSD s proprietary SECTOR Imager and provides high sensitivity and a broad dynamic range The kit includes a plate coated with capture antibodies detection antibodies calibrators controls 10X Read Buffer 10X Wash Buffer and Blocker A It allows for the quantitative measurement of TGF-beta 2 levels with accuracy and reproducibility This assay kit enables efficient and reliable biomarker analysis for preclinical research
Catalog No. 50-000-80742 Supplier Meso Scale Discovery Supplier No. K156XUK24
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX NHP TGF-beta 2 Assay Kit is a multiplex immunoassay that measures active and total TGF-beta 2 levels in non-human primate serum plasma and cell culture supernatants This electrochemiluminescent assay utilizes MSD s proprietary SECTOR Imager and provides high sensitivity and a broad dynamic range The kit includes a plate coated with capture antibodies detection antibodies calibrators controls 10X Read Buffer 10X Wash Buffer and Blocker A It allows for the quantitative measurement of TGF-beta 2 levels with accuracy and reproducibility This assay kit enables efficient and reliable biomarker analysis for preclinical research