Learn More
Meso Scale Discovery UPLEX NHP TGFB3 ASYQP 25 PL
SDP
Catalog No. 5000080874
Encompass
The Meso Scale Discovery U-PLEX NHP TGF-beta3 Assay is a multiplex immunoassay kit that enables simultaneous quantification of active and total TGF-beta3 in non-human primate serum plasma and cell culture supernatants This electrochemiluminescent assay utilizes MSD s proprietary SECTOR Imager and provides superior sensitivity a broad dynamic range and requires low sample volume The kit includes a 96-well plate pre-coated with capture antibodies detection antibodies calibrators controls 10X diluent read buffer and wash buffer sufficient for one 96-well plate This assay allows for efficient and reproducible measurement of TGF-beta3 levels
Catalog No. 50-000-80874 Supplier Meso Scale Discovery Supplier No. K156XVK24
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX NHP TGF-beta3 Assay is a multiplex immunoassay kit that enables simultaneous quantification of active and total TGF-beta3 in non-human primate serum plasma and cell culture supernatants This electrochemiluminescent assay utilizes MSD s proprietary SECTOR Imager and provides superior sensitivity a broad dynamic range and requires low sample volume The kit includes a 96-well plate pre-coated with capture antibodies detection antibodies calibrators controls 10X diluent read buffer and wash buffer sufficient for one 96-well plate This assay allows for efficient and reproducible measurement of TGF-beta3 levels