Learn More
Meso Scale Discovery UPLEX NHP TNF ASY384well
SDP
Catalog No. 5000081163
Encompass
The Meso Scale Discovery U-PLEX NHP TNF-alpha 384-well Assay Kit is a multiplex immunoassay that measures human TNF-alpha in serum plasma and cell culture supernatants It utilizes electrochemiluminescence detection technology and provides high sensitivity broad dynamic range and intra- and inter-assay precision with a streamlined easy-to-use workflow The kit includes a 384-well plate coated with capture antibodies detection antibodies labeled with electrochemiluminescent tags calibrators and assay diluent It allows high-throughput screening and has a typical assay time of 3 5 hours The lower limit of detection is 0 04 pg/mL and the upper limit of detection is 10 000 pg/mL
Catalog No. 50-000-81163 Supplier Meso Scale Discovery Supplier No. K256UCK4
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX NHP TNF-alpha 384-well Assay Kit is a multiplex immunoassay that measures human TNF-alpha in serum plasma and cell culture supernatants It utilizes electrochemiluminescence detection technology and provides high sensitivity broad dynamic range and intra- and inter-assay precision with a streamlined easy-to-use workflow The kit includes a 384-well plate coated with capture antibodies detection antibodies labeled with electrochemiluminescent tags calibrators and assay diluent It allows high-throughput screening and has a typical assay time of 3 5 hours The lower limit of detection is 0 04 pg/mL and the upper limit of detection is 10 000 pg/mL