Learn More
Meso Scale Discovery UPLEX NHP TNFB ASY384well
SDP
Catalog No. 5000078421
Encompass
The Meso Scale Discovery U-PLEX NHP TNF-beta 384-well Assay Kit is a multiplex immunoassay for measuring TNF-beta in human serum plasma and cell culture supernatants It uses electrochemiluminescence detection for high sensitivity and rapid results The kit includes a 384-well plate with capture antibodies detection antibodies calibrators controls buffers and seals It allows simultaneous measurement of TNF-beta with other biomarkers on the U-PLEX platform ensuring high performance and reliability in biomarker quantification for research
Catalog No. 50-000-78421 Supplier Meso Scale Discovery Supplier No. K256UUK4
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX NHP TNF-beta 384-well Assay Kit is a multiplex immunoassay for measuring TNF-beta in human serum plasma and cell culture supernatants It uses electrochemiluminescence detection for high sensitivity and rapid results The kit includes a 384-well plate with capture antibodies detection antibodies calibrators controls buffers and seals It allows simultaneous measurement of TNF-beta with other biomarkers on the U-PLEX platform ensuring high performance and reliability in biomarker quantification for research