Learn More
Meso Scale Discovery UPLEX NHP TPO ASY384well
SDP
Catalog No. 5000080263
Encompass
The Meso Scale Discovery U-PLEX NHP TPO 384-Well Assay is a multiplex immunoassay kit that enables simultaneous quantification of 8 biomarkers related to thyroid peroxidase autoantibodies in human serum plasma and cell culture supernatant samples This 384-well plate assay utilizes MSD s proprietary electroluminescent detection technology and provides high sensitivity up to 5 logs of dynamic range and low sample volume requirements The assay requires 10 L of sample per well and can be read on MSD s SECTOR Imager 6000 The kit includes a plate coated with capture antibodies detection antibodies calibrators and controls
Catalog No. 50-000-80263 Supplier Meso Scale Discovery Supplier No. K256VKK4
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX NHP TPO 384-Well Assay is a multiplex immunoassay kit that enables simultaneous quantification of 8 biomarkers related to thyroid peroxidase autoantibodies in human serum plasma and cell culture supernatant samples This 384-well plate assay utilizes MSD s proprietary electroluminescent detection technology and provides high sensitivity up to 5 logs of dynamic range and low sample volume requirements The assay requires 10 L of sample per well and can be read on MSD s SECTOR Imager 6000 The kit includes a plate coated with capture antibodies detection antibodies calibrators and controls