Learn More
Meso Scale Discovery UPLEX NHP TRAIL ASY384well
SDP
Catalog No. 5000080311
Encompass
The U-PLEX NHP Trail Assay Kit from Meso Scale Diagnostics is a multiplex immunoassay for measuring neurofilament light chain (NfL) glial fibrillary acidic protein (GFAP) total tau (t-tau) and phosphorylated tau (p-tau) in human serum and plasma It uses electrochemiluminescence (ECL) for high sensitivity and rapid results The kit includes a 96-well plate antibodies calibrators controls diluent buffer and seals for one plate Recommended sample is 50 L of serum or plasma Testing time is 3 5 hours with 4-day stability at 2-8 C
Catalog No. 50-000-80311 Supplier Meso Scale Discovery Supplier No. K256XTK4
Add to Cart
Add to Cart
Request a Quote
The U-PLEX NHP Trail Assay Kit from Meso Scale Diagnostics is a multiplex immunoassay for measuring neurofilament light chain (NfL) glial fibrillary acidic protein (GFAP) total tau (t-tau) and phosphorylated tau (p-tau) in human serum and plasma It uses electrochemiluminescence (ECL) for high sensitivity and rapid results The kit includes a 96-well plate antibodies calibrators controls diluent buffer and seals for one plate Recommended sample is 50 L of serum or plasma Testing time is 3 5 hours with 4-day stability at 2-8 C