Learn More
Meso Scale Discovery UPLEX NHP VEGFa ASY384well
SDP
Catalog No. 5000080224
Encompass
The Meso Scale Discovery U-PLEX NHP VEGF-A 384-Well Assay Kit is a multiplex immunoassay that measures VEGF-A levels in human serum plasma and cell culture supernatants It utilizes electrochemiluminescent detection technology and provides high sensitivity broad dynamic range and requires low sample volume The kit includes a 384-well plate coated with capture antibodies detection antibodies labeled with electrochemiluminescent tags calibrators and diluent It allows quantification of VEGF-A levels with a lower limit of detection of 0 31 pg/mL and upper limit of 10 000 pg/mL The assay takes approximately 3 hours to complete
Catalog No. 50-000-80224 Supplier Meso Scale Discovery Supplier No. K256UVK4
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX NHP VEGF-A 384-Well Assay Kit is a multiplex immunoassay that measures VEGF-A levels in human serum plasma and cell culture supernatants It utilizes electrochemiluminescent detection technology and provides high sensitivity broad dynamic range and requires low sample volume The kit includes a 384-well plate coated with capture antibodies detection antibodies labeled with electrochemiluminescent tags calibrators and diluent It allows quantification of VEGF-A levels with a lower limit of detection of 0 31 pg/mL and upper limit of 10 000 pg/mL The assay takes approximately 3 hours to complete