Learn More
Meso Scale Discovery UPLEX Obesity Combo 1 ms
SDP
Catalog No. 5000080334
Encompass
The U-PLEX Obesity Combo 1 Mouse Kit from Meso Scale Discovery is a multiplex immunoassay kit for the quantitative measurement of leptin insulin PAI-1 active and resistin in mouse serum plasma and cell culture supernatants The kit contains a 96-well plate coated with capture antibodies detection antibodies Read Buffer Diluent 2 Calibrator Blend and Wash Buffer The electrochemiluminescent assay allows sensitive and accurate quantification of the analytes using an MSD instrument The kit has a testing range of 24 4-20 000 pg/mL for leptin 78-50 000 pg/mL for insulin 1 22-800 pg/mL for PAI-1 and 15 6-10 000 pg/mL for resistin
Catalog No. 50-000-80334 Supplier Meso Scale Discovery Supplier No. K15300K4
Add to Cart
Add to Cart
Request a Quote
The U-PLEX Obesity Combo 1 Mouse Kit from Meso Scale Discovery is a multiplex immunoassay kit for the quantitative measurement of leptin insulin PAI-1 active and resistin in mouse serum plasma and cell culture supernatants The kit contains a 96-well plate coated with capture antibodies detection antibodies Read Buffer Diluent 2 Calibrator Blend and Wash Buffer The electrochemiluminescent assay allows sensitive and accurate quantification of the analytes using an MSD instrument The kit has a testing range of 24 4-20 000 pg/mL for leptin 78-50 000 pg/mL for insulin 1 22-800 pg/mL for PAI-1 and 15 6-10 000 pg/mL for resistin