Learn More
Meso Scale Discovery UPLEX Obesity Combo 1 rt
SDP
Catalog No. 5000081875
Encompass
The Meso Scale Discovery U-PLEX Obesity Combo 1 Rat Kit is a multiplex immunoassay that simultaneously measures biomarkers related to obesity and metabolic syndrome in rat samples The kit includes a 96-well plate coated with capture antibodies for leptin insulin adiponectin and resistin It utilizes electrochemiluminescent detection for high sensitivity and broad dynamic range The kit includes all necessary reagents and can process up to 80 samples in duplicate It is designed for research use only to study obesity and metabolic disease mechanisms in rat models The kit must be used with Meso Scale Discovery instrumentation and software for detection and data analysis
Catalog No. 50-000-81875 Supplier Meso Scale Discovery Supplier No. K15310K4
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX Obesity Combo 1 Rat Kit is a multiplex immunoassay that simultaneously measures biomarkers related to obesity and metabolic syndrome in rat samples The kit includes a 96-well plate coated with capture antibodies for leptin insulin adiponectin and resistin It utilizes electrochemiluminescent detection for high sensitivity and broad dynamic range The kit includes all necessary reagents and can process up to 80 samples in duplicate It is designed for research use only to study obesity and metabolic disease mechanisms in rat models The kit must be used with Meso Scale Discovery instrumentation and software for detection and data analysis