Learn More
Meso Scale Discovery UPLEX Obesity Combo 2 ms
SDP
Catalog No. 5000082491
Encompass
The U-PLEX Obesity Combo 2 Mouse is a multiplex assay kit by Meso Scale Diagnostics that allows simultaneous measurement of leptin insulin PAI-1 active and resistin in mouse samples It utilizes electrochemiluminescent detection technology for high sensitivity and broad dynamic range The kit includes a 96-well plate coated with capture antibodies detection antibodies read buffer wash buffer diluent and calibrators It is compatible with serum plasma and tissue culture supernatant samples The assay takes approximately 3 5 hours to complete after a 2 hour incubation
Catalog No. 50-000-82491 Supplier Meso Scale Discovery Supplier No. K15301K4
Add to Cart
Add to Cart
Request a Quote
The U-PLEX Obesity Combo 2 Mouse is a multiplex assay kit by Meso Scale Diagnostics that allows simultaneous measurement of leptin insulin PAI-1 active and resistin in mouse samples It utilizes electrochemiluminescent detection technology for high sensitivity and broad dynamic range The kit includes a 96-well plate coated with capture antibodies detection antibodies read buffer wash buffer diluent and calibrators It is compatible with serum plasma and tissue culture supernatant samples The assay takes approximately 3 5 hours to complete after a 2 hour incubation