Learn More
Meso Scale Discovery UPLEX Obesity Combo 2 rt
SDP
Catalog No. 5000082128
Encompass
The Meso Scale Discovery U-PLEX Obesity Combo 2 Rat Kit is a multiplex immunoassay that simultaneously measures the levels of leptin insulin and C-peptide in rat serum plasma and tissue culture supernatants This assay kit utilizes MSD s electrochemiluminescent detection technology and provides high sensitivity up to 5 logs of dynamic range and low sample volume requirements The kit includes a 96-well plate pre-coated with capture antibodies detection antibodies and calibrators to generate standard curves This assay allows for the efficient analysis of obesity and metabolic biomarker levels in rat samples
Catalog No. 50-000-82128 Supplier Meso Scale Discovery Supplier No. K15311K4
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX Obesity Combo 2 Rat Kit is a multiplex immunoassay that simultaneously measures the levels of leptin insulin and C-peptide in rat serum plasma and tissue culture supernatants This assay kit utilizes MSD s electrochemiluminescent detection technology and provides high sensitivity up to 5 logs of dynamic range and low sample volume requirements The kit includes a 96-well plate pre-coated with capture antibodies detection antibodies and calibrators to generate standard curves This assay allows for the efficient analysis of obesity and metabolic biomarker levels in rat samples