Learn More
Meso Scale Discovery UPLEX Plus HUM Synuclein
SDP
Catalog No. 5000081479
Encompass
The Meso Scale Discovery U-PLEX Plus Human Alpha-Synuclein Kit is a multiplex immunoassay for measuring alpha-synuclein in human serum plasma and CSF It uses electrochemiluminescence for high sensitivity and rapid results The kit includes a 96-well plate detection antibodies calibrators controls diluent buffers and seals Sensitivity is 0 064 pg/mL with a quantification limit of 2500 pg/mL The assay takes 3 5 hours aiding in understanding synuclein biology and pathology
Catalog No. 50-000-81479 Supplier Meso Scale Discovery Supplier No. K151WKP4
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX Plus Human Alpha-Synuclein Kit is a multiplex immunoassay for measuring alpha-synuclein in human serum plasma and CSF It uses electrochemiluminescence for high sensitivity and rapid results The kit includes a 96-well plate detection antibodies calibrators controls diluent buffers and seals Sensitivity is 0 064 pg/mL with a quantification limit of 2500 pg/mL The assay takes 3 5 hours aiding in understanding synuclein biology and pathology