Learn More
Meso Scale Discovery UPLEX Proinflam Combo 1 NHP
SDP
Catalog No. 5000081698
Encompass
The U-PLEX ProInflam Combo 1 NHP is a multiplex assay kit by Meso Scale Diagnostics that allows simultaneous quantitative measurement of 10 human proinflammatory biomarkers The assay utilizes electrochemiluminescent detection technology and provides high sensitivity broad dynamic range and low sample volume requirements This kit includes reagents for analyzing IL-1beta IL-6 IL-8 TNF-alpha IFN-gamma IL-10 IL-12p70 IL-17A/F IL-18 and IL-23 in cell culture supernatants serum plasma and other biological samples The kit contains a 96-well plate coated with capture antibodies detection antibodies read buffer wash buffer calibrator blend and serum and plasma diluents
Catalog No. 50-000-81698 Supplier Meso Scale Discovery Supplier No. K15070K4
Add to Cart
Add to Cart
Request a Quote
The U-PLEX ProInflam Combo 1 NHP is a multiplex assay kit by Meso Scale Diagnostics that allows simultaneous quantitative measurement of 10 human proinflammatory biomarkers The assay utilizes electrochemiluminescent detection technology and provides high sensitivity broad dynamic range and low sample volume requirements This kit includes reagents for analyzing IL-1beta IL-6 IL-8 TNF-alpha IFN-gamma IL-10 IL-12p70 IL-17A/F IL-18 and IL-23 in cell culture supernatants serum plasma and other biological samples The kit contains a 96-well plate coated with capture antibodies detection antibodies read buffer wash buffer calibrator blend and serum and plasma diluents