Learn More
Meso Scale Discovery UPLEX Proinflam Combo 2 hu
SDP
Catalog No. 5000080570
Encompass
The Meso Scale Discovery U-PLEX ProInflam Combo 2 Human Kit is a multiplex immunoassay that simultaneously measures human IFN-gamma IL-10 IL-12p70 IL-13 IL-1beta IL-2 IL-4 IL-6 IL-8 TNF-alpha in serum plasma cell culture supernatants and other biological fluids This assay utilizes MSD s electrochemiluminescent detection technology and is designed for use on MSD s SECTOR Imager 6000 The kit includes a 96-well plate coated with capture antibodies detection antibodies calibrators controls 10X Read Buffer 10X Wash Buffer and Plate Seals This assay allows for the rapid sensitive and accurate measurement of multiple proinflammatory cytokines from a small sample volume
Catalog No. 50-000-80570 Supplier Meso Scale Discovery Supplier No. K15066K4
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX ProInflam Combo 2 Human Kit is a multiplex immunoassay that simultaneously measures human IFN-gamma IL-10 IL-12p70 IL-13 IL-1beta IL-2 IL-4 IL-6 IL-8 TNF-alpha in serum plasma cell culture supernatants and other biological fluids This assay utilizes MSD s electrochemiluminescent detection technology and is designed for use on MSD s SECTOR Imager 6000 The kit includes a 96-well plate coated with capture antibodies detection antibodies calibrators controls 10X Read Buffer 10X Wash Buffer and Plate Seals This assay allows for the rapid sensitive and accurate measurement of multiple proinflammatory cytokines from a small sample volume