Learn More
Meso Scale Discovery UPLEX Proinflam Combo 3 hu
SDP
Catalog No. 5000079191
Encompass
The Meso Scale Discovery U-PLEX ProInflam Combo 3 Human Kit is a multiplex immunoassay that simultaneously measures IL-6 IL-8 and TNF-alpha in human serum plasma and cell culture supernatants It utilizes electrochemiluminescence detection technology and provides high sensitivity broad dynamic range and intra- and inter-assay precision The kit includes a 96-well plate coated with capture antibodies detection antibodies labeled with MSD SULFO-TAG Read Buffer 10X Diluent and calibrators and controls to generate a standard curve and validate assay performance It allows efficient and accurate quantification of multiple inflammatory biomarkers from a small sample volume
Catalog No. 50-000-79191 Supplier Meso Scale Discovery Supplier No. K15052K4
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX ProInflam Combo 3 Human Kit is a multiplex immunoassay that simultaneously measures IL-6 IL-8 and TNF-alpha in human serum plasma and cell culture supernatants It utilizes electrochemiluminescence detection technology and provides high sensitivity broad dynamic range and intra- and inter-assay precision The kit includes a 96-well plate coated with capture antibodies detection antibodies labeled with MSD SULFO-TAG Read Buffer 10X Diluent and calibrators and controls to generate a standard curve and validate assay performance It allows efficient and accurate quantification of multiple inflammatory biomarkers from a small sample volume