Learn More
Meso Scale Discovery UPLEX Proinflam Combo 4 hu
SDP
Catalog No. 5000078810
Encompass
The Meso Scale Discovery U-PLEX ProInflam Combo 4 Human Kit is a multiplex immunoassay that simultaneously measures the levels of four human proinflammatory biomarkers IFN- IL-1 IL-6 and TNF- in serum plasma and cell culture supernatants This kit utilizes MSD s electrochemiluminescent detection technology and provides high sensitivity up to 5-logs of dynamic range and low sample volume requirements The kit includes a 96-well plate coated with capture antibodies detection antibodies Read Buffer Diluent 2 Calibrator Blend and Wash Buffer This assay can be used for biomarker profiling mechanistic studies and pharmacodynamic monitoring of proinflammatory diseases
Catalog No. 50-000-78810 Supplier Meso Scale Discovery Supplier No. K15053K4
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX ProInflam Combo 4 Human Kit is a multiplex immunoassay that simultaneously measures the levels of four human proinflammatory biomarkers IFN- IL-1 IL-6 and TNF- in serum plasma and cell culture supernatants This kit utilizes MSD s electrochemiluminescent detection technology and provides high sensitivity up to 5-logs of dynamic range and low sample volume requirements The kit includes a 96-well plate coated with capture antibodies detection antibodies Read Buffer Diluent 2 Calibrator Blend and Wash Buffer This assay can be used for biomarker profiling mechanistic studies and pharmacodynamic monitoring of proinflammatory diseases