Learn More
Meso Scale Discovery UPLEX Rat BDNF ASY384well
SDP
Catalog No. 5000081161
Encompass
The Meso Scale Discovery U-PLEX Rat BDNF 384-well Assay Kit is a multiplex immunoassay that measures brain-derived neurotrophic factor (BDNF) in rat serum plasma and cell culture supernatants The kit uses electrochemiluminescence (ECL) detection technology and provides a 384-well plate formatted assay for high throughput analysis of BDNF with high sensitivity and broad dynamic range This quantitative immunoassay allows users to accurately determine BDNF levels with as little as 25 L of sample The kit includes a 384-well MULTI-ARRAY Rat BDNF Assay Plate Detection Antibody Read Buffer Calibrator Diluent and SULFO-TAG labeled anti-rat BDNF Antibody
Catalog No. 50-000-81161 Supplier Meso Scale Discovery Supplier No. K2536WK4
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX Rat BDNF 384-well Assay Kit is a multiplex immunoassay that measures brain-derived neurotrophic factor (BDNF) in rat serum plasma and cell culture supernatants The kit uses electrochemiluminescence (ECL) detection technology and provides a 384-well plate formatted assay for high throughput analysis of BDNF with high sensitivity and broad dynamic range This quantitative immunoassay allows users to accurately determine BDNF levels with as little as 25 L of sample The kit includes a 384-well MULTI-ARRAY Rat BDNF Assay Plate Detection Antibody Read Buffer Calibrator Diluent and SULFO-TAG labeled anti-rat BDNF Antibody