Learn More
Meso Scale Discovery UPLEX Rat CPeptide ASY384well
SDP
Catalog No. 5000080002
Encompass
The Meso Scale Discovery U-PLEX Rat C-Peptide 384-Well Assay is an electrochemiluminescent immunoassay for the quantitative measurement of C-peptide in rat serum plasma and other biological samples This 10-plate kit includes a 384-well plate coated with capture antibody detection antibody conjugated to an electrochemiluminescent label calibrators and diluent The assay has a typical sensitivity of 0 008 ng/mL and an upper limit of quantification of 50 ng/mL This product allows high-throughput analysis of C-peptide levels with low sample volume requirements
Catalog No. 50-000-80002 Supplier Meso Scale Discovery Supplier No. K2536JK4
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX Rat C-Peptide 384-Well Assay is an electrochemiluminescent immunoassay for the quantitative measurement of C-peptide in rat serum plasma and other biological samples This 10-plate kit includes a 384-well plate coated with capture antibody detection antibody conjugated to an electrochemiluminescent label calibrators and diluent The assay has a typical sensitivity of 0 008 ng/mL and an upper limit of quantification of 50 ng/mL This product allows high-throughput analysis of C-peptide levels with low sample volume requirements