Learn More
Meso Scale Discovery UPLEX Rat CPeptide ASYQP
SDP
Catalog No. 5000082185
Encompass
The Meso Scale Discovery U-PLEX Rat C-Peptide Assay Kit is a multiplex immunoassay for quantifying C-peptide in rat serum plasma and other samples It uses MSD s electrochemiluminescent detection technology offering high sensitivity a wide dynamic range and low sample volume needs The kit includes a 96-well plate with capture antibodies detection antibodies and necessary calibrators and controls Results are available in 3 5 hours using an MSD instrument like SECTOR Imager or QuickPlex enabling accurate C-peptide measurement for research
Catalog No. 50-000-82185 Supplier Meso Scale Discovery Supplier No. K1536JK24
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX Rat C-Peptide Assay Kit is a multiplex immunoassay for quantifying C-peptide in rat serum plasma and other samples It uses MSD s electrochemiluminescent detection technology offering high sensitivity a wide dynamic range and low sample volume needs The kit includes a 96-well plate with capture antibodies detection antibodies and necessary calibrators and controls Results are available in 3 5 hours using an MSD instrument like SECTOR Imager or QuickPlex enabling accurate C-peptide measurement for research