Learn More
Meso Scale Discovery UPLEX Rat Ghrelin act ASY384
SDP
Catalog No. 5000081221
Encompass
The Meso Scale Discovery U-PLEX Rat Ghrelin (Active) 384-Well Assay Kit is a multiplex immunoassay that measures active ghrelin in rat serum plasma and cell culture supernatants This kit uses electrochemiluminescence detection technology and provides a 384-well plate formatted assay allowing high throughput screening The kit includes a plate coated with capture antibodies detection antibodies labeled with an electrochemiluminescent label and calibrator containing active rat ghrelin This assay has high sensitivity wide dynamic range and requires low sample volume Overall this is a high-performance assay for quantifying active ghrelin levels in rat biological samples
Catalog No. 50-000-81221 Supplier Meso Scale Discovery Supplier No. K2536KK2
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX Rat Ghrelin (Active) 384-Well Assay Kit is a multiplex immunoassay that measures active ghrelin in rat serum plasma and cell culture supernatants This kit uses electrochemiluminescence detection technology and provides a 384-well plate formatted assay allowing high throughput screening The kit includes a plate coated with capture antibodies detection antibodies labeled with an electrochemiluminescent label and calibrator containing active rat ghrelin This assay has high sensitivity wide dynamic range and requires low sample volume Overall this is a high-performance assay for quantifying active ghrelin levels in rat biological samples