Learn More
Meso Scale Discovery UPLEX Rat Ghrelin act ASY384
SDP
Catalog No. 5000081288
Encompass
The Meso Scale Discovery U-PLEX Rat Ghrelin (Active) 384-Well Assay is a multiplex immunoassay kit that measures active ghrelin levels in rat serum plasma and cell culture supernatants It uses electrochemiluminescence detection technology and provides a sensitive accurate and reproducible way to quantify active ghrelin levels from small sample volumes The kit includes a 384-well plate coated with capture antibodies detection antibodies calibrators controls 10X diluent read buffer and wash buffer sufficient for performing 384 tests in duplicate It has a detection range of 50-50 000 pg/mL and requires just 25 L of sample per well
Catalog No. 50-000-81288 Supplier Meso Scale Discovery Supplier No. K2536KK4
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX Rat Ghrelin (Active) 384-Well Assay is a multiplex immunoassay kit that measures active ghrelin levels in rat serum plasma and cell culture supernatants It uses electrochemiluminescence detection technology and provides a sensitive accurate and reproducible way to quantify active ghrelin levels from small sample volumes The kit includes a 384-well plate coated with capture antibodies detection antibodies calibrators controls 10X diluent read buffer and wash buffer sufficient for performing 384 tests in duplicate It has a detection range of 50-50 000 pg/mL and requires just 25 L of sample per well