Learn More
Meso Scale Discovery UPLEX Rat Ghrelin active ASY
SDP
Catalog No. 5000078585
Encompass
The Meso Scale Discovery U-PLEX Rat Ghrelin (Active) Assay Kit is a multiplex immunoassay that measures active ghrelin in rat serum plasma and cell culture supernatants This assay uses electrochemiluminescence (ECL) detection technology and provides high sensitivity broad dynamic range and intra- and inter-assay precision The kit includes a 96-well plate coated with capture antibodies detection antibodies labeled with ruthenium and calibrators and controls It allows simultaneous measurement of active ghrelin and SOD1 in the same sample for internal assay quality control The assay requires 50 L of sample and can be read on MSD instrument platforms
Catalog No. 50-000-78585 Supplier Meso Scale Discovery Supplier No. K1536KK1
The Meso Scale Discovery U-PLEX Rat Ghrelin (Active) Assay Kit is a multiplex immunoassay that measures active ghrelin in rat serum plasma and cell culture supernatants This assay uses electrochemiluminescence (ECL) detection technology and provides high sensitivity broad dynamic range and intra- and inter-assay precision The kit includes a 96-well plate coated with capture antibodies detection antibodies labeled with ruthenium and calibrators and controls It allows simultaneous measurement of active ghrelin and SOD1 in the same sample for internal assay quality control The assay requires 50 L of sample and can be read on MSD instrument platforms