Learn More
Meso Scale Discovery UPLEX Rat Ghrelin active ASY
SDP
Catalog No. 5000081915
Encompass
The Meso Scale Discovery U-PLEX Rat Ghrelin (Active) Assay Kit is an electrochemiluminescent immunoassay for the quantitative measurement of active ghrelin in rat serum plasma and cell culture supernatants This assay uses MSD s proprietary SULFO-TAG labels and multi-array technology to provide sensitive and rapid detection of active ghrelin The kit includes a 96-well plate coated with capture antibody detection antibody conjugated to ruthenium label calibrators and sample diluent The assay has a typical sensitivity of 0 3 pg/mL and an upper limit of quantification of 2500 pg/mL This product allows for high-throughput analysis of active ghrelin levels
Catalog No. 50-000-81915 Supplier Meso Scale Discovery Supplier No. K1536KK2
The Meso Scale Discovery U-PLEX Rat Ghrelin (Active) Assay Kit is an electrochemiluminescent immunoassay for the quantitative measurement of active ghrelin in rat serum plasma and cell culture supernatants This assay uses MSD s proprietary SULFO-TAG labels and multi-array technology to provide sensitive and rapid detection of active ghrelin The kit includes a 96-well plate coated with capture antibody detection antibody conjugated to ruthenium label calibrators and sample diluent The assay has a typical sensitivity of 0 3 pg/mL and an upper limit of quantification of 2500 pg/mL This product allows for high-throughput analysis of active ghrelin levels