Learn More
Meso Scale Discovery UPLEX Rat Ghrelin active ASYQP
SDP
Catalog No. 5000079761
Encompass
The Meso Scale Discovery U-PLEX Rat Ghrelin (Active) Assay Kit is a multiplex immunoassay that measures active ghrelin in rat serum plasma and cell culture supernatants This assay uses electrochemiluminescence (ECL) detection technology and provides high sensitivity broad dynamic range and intra- and inter-assay precision The kit includes a 96-well plate coated with capture antibodies detection antibodies labeled with an ECL tag calibrators and controls The active ghrelin assay has a sensitivity of 0 3 pg/mL and requires 25 L sample size This quantitative immunoassay allows simultaneous detection of multiple biomarkers using MSD s proprietary QuickPlex platform
Catalog No. 50-000-79761 Supplier Meso Scale Discovery Supplier No. K1536KK21
The Meso Scale Discovery U-PLEX Rat Ghrelin (Active) Assay Kit is a multiplex immunoassay that measures active ghrelin in rat serum plasma and cell culture supernatants This assay uses electrochemiluminescence (ECL) detection technology and provides high sensitivity broad dynamic range and intra- and inter-assay precision The kit includes a 96-well plate coated with capture antibodies detection antibodies labeled with an ECL tag calibrators and controls The active ghrelin assay has a sensitivity of 0 3 pg/mL and requires 25 L sample size This quantitative immunoassay allows simultaneous detection of multiple biomarkers using MSD s proprietary QuickPlex platform