Learn More
Meso Scale Discovery UPLEX Rat Ghrelin TTL ASY
SDP
Catalog No. 5000080347
Encompass
The Meso Scale Discovery U-PLEX Rat Ghrelin Total Assay Kit is an electrochemiluminescent immunoassay for measuring total ghrelin in rat serum plasma and tissue culture media It allows simultaneous measurement of ghrelin and other biomarkers using MSD s SULFO-TAG labels and multi-array technology The kit includes a 96-well plate with capture antibodies detection antibodies labeled with ruthenium read buffer wash buffer blocker solutions calibrator and controls The assay sensitivity is 0 3 pg/mL with an upper quantification limit of 2500 pg/mL
Catalog No. 50-000-80347 Supplier Meso Scale Discovery Supplier No. K1535XK4
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX Rat Ghrelin Total Assay Kit is an electrochemiluminescent immunoassay for measuring total ghrelin in rat serum plasma and tissue culture media It allows simultaneous measurement of ghrelin and other biomarkers using MSD s SULFO-TAG labels and multi-array technology The kit includes a 96-well plate with capture antibodies detection antibodies labeled with ruthenium read buffer wash buffer blocker solutions calibrator and controls The assay sensitivity is 0 3 pg/mL with an upper quantification limit of 2500 pg/mL