Learn More
Meso Scale Discovery UPLEX Rat Ghrelin TTL ASY384
SDP
Catalog No. 5000079341
Encompass
The Meso Scale Discovery U-PLEX Rat Ghrelin Total 384-Well Assay Kit is a multiplex immunoassay for the quantitative measurement of total ghrelin in rat serum plasma and cell culture supernatants The kit contains a 384-well plate coated with capture antibodies against total ghrelin detection antibodies labeled with an electrochemiluminescent label calibrator solutions assay diluent read buffer wash buffer and plate seals The assay has a typical sensitivity of 0 3 pg/mL and an upper limit of quantification of 10 000 pg/mL for total ghrelin The kit includes sufficient reagents for performing 384 assays in a 384-well plate using a small sample volume of 25 L
Catalog No. 50-000-79341 Supplier Meso Scale Discovery Supplier No. K2535XK2
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX Rat Ghrelin Total 384-Well Assay Kit is a multiplex immunoassay for the quantitative measurement of total ghrelin in rat serum plasma and cell culture supernatants The kit contains a 384-well plate coated with capture antibodies against total ghrelin detection antibodies labeled with an electrochemiluminescent label calibrator solutions assay diluent read buffer wash buffer and plate seals The assay has a typical sensitivity of 0 3 pg/mL and an upper limit of quantification of 10 000 pg/mL for total ghrelin The kit includes sufficient reagents for performing 384 assays in a 384-well plate using a small sample volume of 25 L