Learn More
Meso Scale Discovery UPLEX Rat Ghrelin TTL ASY384
SDP
Catalog No. 5000081236
Encompass
The Meso Scale Discovery U-PLEX Rat Ghrelin Total 384-Well Assay Kit is a multiplex immunoassay that measures total ghrelin levels in rat serum plasma and cell culture supernatants This kit uses electrochemiluminescence detection technology and provides a 384-well plate formatted assay allowing high throughput screening The assay has a sensitivity of 0 3 pg/mL and a broad dynamic range of over 3 logs Each kit provides reagents for one 384-well plate including antibody-coated plates detection antibodies calibrators and controls This assay allows for the accurate sensitive and rapid measurement of total ghrelin levels from rat samples
Catalog No. 50-000-81236 Supplier Meso Scale Discovery Supplier No. K2535XK4
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX Rat Ghrelin Total 384-Well Assay Kit is a multiplex immunoassay that measures total ghrelin levels in rat serum plasma and cell culture supernatants This kit uses electrochemiluminescence detection technology and provides a 384-well plate formatted assay allowing high throughput screening The assay has a sensitivity of 0 3 pg/mL and a broad dynamic range of over 3 logs Each kit provides reagents for one 384-well plate including antibody-coated plates detection antibodies calibrators and controls This assay allows for the accurate sensitive and rapid measurement of total ghrelin levels from rat samples