Learn More
Meso Scale Discovery UPLEX Rat Ghrelin TTL ASYQP
SDP
Catalog No. 5000079762
Encompass
The Meso Scale Discovery U-PLEX Rat Ghrelin Total Assay Kit is a multiplex immunoassay for measuring total ghrelin in rat serum plasma and cell culture supernatants It uses MSD s electrochemiluminescent detection technology offering high sensitivity a wide dynamic range and low sample volume needs The kit includes a 96-well plate with capture antibodies detection antibodies calibrators and controls Results are read on MSD s SECTOR Imager and analyzed with MSD Discovery Workbench software aiding in the study of appetite regulation and energy metabolism in rat models
Catalog No. 50-000-79762 Supplier Meso Scale Discovery Supplier No. K1535XK24
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX Rat Ghrelin Total Assay Kit is a multiplex immunoassay for measuring total ghrelin in rat serum plasma and cell culture supernatants It uses MSD s electrochemiluminescent detection technology offering high sensitivity a wide dynamic range and low sample volume needs The kit includes a 96-well plate with capture antibodies detection antibodies calibrators and controls Results are read on MSD s SECTOR Imager and analyzed with MSD Discovery Workbench software aiding in the study of appetite regulation and energy metabolism in rat models