Learn More
Meso Scale Discovery UPLEX Rat Ghrelin TTL ASYQP
SDP
Catalog No. 5000079796
Encompass
The Meso Scale Discovery U-PLEX Rat Ghrelin Total Assay Kit is a multiplex immunoassay that measures total ghrelin levels in rat serum plasma and cell culture supernatants This assay uses electrochemiluminescence (ECL) detection technology and provides high sensitivity broad dynamic range and intra- and inter-assay precision The kit includes a 96-well plate pre-coated with capture antibodies detection antibodies calibrators controls 10X diluent read buffer and wash buffer The total time to run the assay is around 3 5 hours This kit allows for the quantitative measurement of total ghrelin levels in rat biological samples
Catalog No. 50-000-79796 Supplier Meso Scale Discovery Supplier No. K1535XK21
The Meso Scale Discovery U-PLEX Rat Ghrelin Total Assay Kit is a multiplex immunoassay that measures total ghrelin levels in rat serum plasma and cell culture supernatants This assay uses electrochemiluminescence (ECL) detection technology and provides high sensitivity broad dynamic range and intra- and inter-assay precision The kit includes a 96-well plate pre-coated with capture antibodies detection antibodies calibrators controls 10X diluent read buffer and wash buffer The total time to run the assay is around 3 5 hours This kit allows for the quantitative measurement of total ghrelin levels in rat biological samples