Learn More
Meso Scale Discovery UPLEX Rat Ghrelin TTL ASYQP
SDP
Catalog No. 5000082188
Encompass
The Meso Scale Discovery U-PLEX Rat Ghrelin Total Assay Kit is a multiplex immunoassay that measures total ghrelin levels in rat serum plasma and cell culture supernatants This assay uses electrochemiluminescence (ECL) detection technology and provides high sensitivity broad dynamic range and intra- and inter-assay precision The kit includes a 96-well plate coated with capture antibodies detection antibodies labeled with ruthenium calibrators and controls Results can be read on the MSD QuickPlex SQ 120 instrument and data analyzed using MSD Discovery Workbench software This assay allows rapid and accurate quantification of total ghrelin levels from rat biological samples
Catalog No. 50-000-82188 Supplier Meso Scale Discovery Supplier No. K1535XK22
The Meso Scale Discovery U-PLEX Rat Ghrelin Total Assay Kit is a multiplex immunoassay that measures total ghrelin levels in rat serum plasma and cell culture supernatants This assay uses electrochemiluminescence (ECL) detection technology and provides high sensitivity broad dynamic range and intra- and inter-assay precision The kit includes a 96-well plate coated with capture antibodies detection antibodies labeled with ruthenium calibrators and controls Results can be read on the MSD QuickPlex SQ 120 instrument and data analyzed using MSD Discovery Workbench software This assay allows rapid and accurate quantification of total ghrelin levels from rat biological samples