Learn More
Meso Scale Discovery UPLEX Rat GLP1 active ASY
SDP
Catalog No. 5000081965
Encompass
The Meso Scale Discovery U-PLEX Rat GLP-1 (Active) Assay Kit is a multiplex immunoassay that measures active GLP-1 in rat serum plasma and cell culture supernatants This electrochemiluminescent assay utilizes MSD s proprietary SULFO-TAG labels and multi-array technology to provide sensitive and rapid quantification of GLP-1 levels with low sample volume requirements The kit includes a 96-well plate pre-coated with capture antibodies detection antibodies calibrators controls 10X Read Buffer 10X Wash Buffer and Plate Seals The assay has a detection range of 1 23-2500 pg/mL and includes reagents for one 96-well plate
Catalog No. 50-000-81965 Supplier Meso Scale Discovery Supplier No. K1536LK4
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX Rat GLP-1 (Active) Assay Kit is a multiplex immunoassay that measures active GLP-1 in rat serum plasma and cell culture supernatants This electrochemiluminescent assay utilizes MSD s proprietary SULFO-TAG labels and multi-array technology to provide sensitive and rapid quantification of GLP-1 levels with low sample volume requirements The kit includes a 96-well plate pre-coated with capture antibodies detection antibodies calibrators controls 10X Read Buffer 10X Wash Buffer and Plate Seals The assay has a detection range of 1 23-2500 pg/mL and includes reagents for one 96-well plate