Learn More
Meso Scale Discovery UPLEX Rat GLP1 active ASYQP
SDP
Catalog No. 5000081365
Encompass
The Meso Scale Discovery U-PLEX Rat GLP-1 (Active) Assay Kit is an electrochemiluminescent immunoassay for the quantitative measurement of active GLP-1 in rat serum plasma and cell culture supernatants The assay uses MSD GOLD plate technology coated with a GLP-1 specific capture antibody and a SULFO-TAG labeled detection antibody providing high sensitivity and a broad dynamic range The kit includes a 96-well plate detection antibody sample diluent calibrator and wash buffer for performing the immunoassay on the MESO QuickPlex SQ 120 instrument The assay has high specificity for active GLP-1 and requires a low sample volume of 25 L
Catalog No. 50-000-81365 Supplier Meso Scale Discovery Supplier No. K1536LK24
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX Rat GLP-1 (Active) Assay Kit is an electrochemiluminescent immunoassay for the quantitative measurement of active GLP-1 in rat serum plasma and cell culture supernatants The assay uses MSD GOLD plate technology coated with a GLP-1 specific capture antibody and a SULFO-TAG labeled detection antibody providing high sensitivity and a broad dynamic range The kit includes a 96-well plate detection antibody sample diluent calibrator and wash buffer for performing the immunoassay on the MESO QuickPlex SQ 120 instrument The assay has high specificity for active GLP-1 and requires a low sample volume of 25 L