Learn More
Meso Scale Discovery UPLEX Rat GLP1 active ASYQP
SDP
Catalog No. 5000082078
Encompass
The Meso Scale Discovery U-PLEX Rat GLP-1 (Active) Assay Kit is an electrochemiluminescent immunoassay for the quantitative measurement of active glucagon-like peptide-1 (GLP-1) in rat serum plasma and cell culture supernatants The assay uses MSD GOLD SULFO-TAG labels and is designed for use on the MESO QuickPlex SQ 120 instrument The kit includes a 96-well plate coated with anti-GLP-1 capture antibody detection antibody calibrator and assay diluent The assay has a typical sensitivity of 0 3 pg/mL and an upper limit of quantification of 2500 pg/mL The total assay time is approximately 3 5 hours This product allows for the high-sensitivity quantification of active GLP-1
Catalog No. 50-000-82078 Supplier Meso Scale Discovery Supplier No. K1536LK22
The Meso Scale Discovery U-PLEX Rat GLP-1 (Active) Assay Kit is an electrochemiluminescent immunoassay for the quantitative measurement of active glucagon-like peptide-1 (GLP-1) in rat serum plasma and cell culture supernatants The assay uses MSD GOLD SULFO-TAG labels and is designed for use on the MESO QuickPlex SQ 120 instrument The kit includes a 96-well plate coated with anti-GLP-1 capture antibody detection antibody calibrator and assay diluent The assay has a typical sensitivity of 0 3 pg/mL and an upper limit of quantification of 2500 pg/mL The total assay time is approximately 3 5 hours This product allows for the high-sensitivity quantification of active GLP-1