Learn More
Meso Scale Discovery UPLEX Rat GLP1 inactive ASY
SDP
Catalog No. 5000082213
Encompass
The Meso Scale Discovery U-PLEX Rat GLP-1 (Inactive) Assay Kit is a multiplex immunoassay that measures GLP-1 (7-36) amide and GLP-1 (9-36) amide in rat serum plasma and cell culture supernatants The assay uses electrochemiluminescence detection technology and provides high sensitivity broad dynamic range and rapid results This assay kit includes a 96-well plate coated with capture antibodies detection antibodies labeled with electrochemiluminescent tags calibrators and controls The assay takes approximately 3 5 hours to complete and requires a Meso Scale Discovery instrument for analysis This product allows for the quantitative determination of inactive GLP-1 levels
Catalog No. 50-000-82213 Supplier Meso Scale Discovery Supplier No. K1535VK4
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX Rat GLP-1 (Inactive) Assay Kit is a multiplex immunoassay that measures GLP-1 (7-36) amide and GLP-1 (9-36) amide in rat serum plasma and cell culture supernatants The assay uses electrochemiluminescence detection technology and provides high sensitivity broad dynamic range and rapid results This assay kit includes a 96-well plate coated with capture antibodies detection antibodies labeled with electrochemiluminescent tags calibrators and controls The assay takes approximately 3 5 hours to complete and requires a Meso Scale Discovery instrument for analysis This product allows for the quantitative determination of inactive GLP-1 levels