Learn More
Meso Scale Discovery UPLEX Rat GLP1 inactive ASY
SDP
Catalog No. 5000081506
Encompass
The Meso Scale Discovery U-PLEX Rat GLP-1 (Inactive) Assay Kit K1535VK-2 is an electrochemiluminescent immunoassay for the quantitative measurement of inactive GLP-1 in rat serum plasma and cell culture supernatants The assay uses MSD GOLD SULFO-TAG labels and is designed for use on the MESO QuickPlex SQ 120 instrument The kit includes a 96-well plate coated with capture antibody detection antibody calibrator and sample diluent The assay has a typical sensitivity of 0 3 pg/mL and an upper limit of quantification of 2500 pg/mL
Catalog No. 50-000-81506 Supplier Meso Scale Discovery Supplier No. K1535VK2
The Meso Scale Discovery U-PLEX Rat GLP-1 (Inactive) Assay Kit K1535VK-2 is an electrochemiluminescent immunoassay for the quantitative measurement of inactive GLP-1 in rat serum plasma and cell culture supernatants The assay uses MSD GOLD SULFO-TAG labels and is designed for use on the MESO QuickPlex SQ 120 instrument The kit includes a 96-well plate coated with capture antibody detection antibody calibrator and sample diluent The assay has a typical sensitivity of 0 3 pg/mL and an upper limit of quantification of 2500 pg/mL