Learn More
Meso Scale Discovery UPLEX Rat GLP1 inactive ASYQP
SDP
Catalog No. 5000078593
Encompass
The Meso Scale Discovery U-PLEX Rat GLP-1 (Inactive) Assay Kit measures GLP-1 (7-36) amide and GLP-1 (9-36) amide in rat serum plasma and cell culture supernatants This electrochemiluminescent immunoassay uses MSD s SULFO-TAG labels and multi-array technology for sensitive detection The kit includes a 96-well plate with capture antibodies detection antibodies with ruthenium labels calibrators and necessary reagents Results are read on MSD s SECTOR Imager instruments with a typical assay time of 3 5 hours for 96 samples allowing multiplexed measurement of active and inactive GLP-1
Catalog No. 50-000-78593 Supplier Meso Scale Discovery Supplier No. K1535VK21
The Meso Scale Discovery U-PLEX Rat GLP-1 (Inactive) Assay Kit measures GLP-1 (7-36) amide and GLP-1 (9-36) amide in rat serum plasma and cell culture supernatants This electrochemiluminescent immunoassay uses MSD s SULFO-TAG labels and multi-array technology for sensitive detection The kit includes a 96-well plate with capture antibodies detection antibodies with ruthenium labels calibrators and necessary reagents Results are read on MSD s SECTOR Imager instruments with a typical assay time of 3 5 hours for 96 samples allowing multiplexed measurement of active and inactive GLP-1