Learn More
Meso Scale Discovery UPLEX Rat GLP1 inactive ASYQP
SDP
Catalog No. 5000078594
Encompass
The Meso Scale Discovery U-PLEX Rat GLP-1 (Inactive) Assay Kit is a multiplex immunoassay that measures GLP-1 (7-36) amide and GLP-1 (9-36) amide in rat serum plasma and cell culture supernatants The assay uses electrochemiluminescence detection technology and provides high sensitivity broad dynamic range and rapid results This assay allows for the simultaneous detection of both active and inactive forms of GLP-1 enabling more complete characterization of GLP-1 biology The kit includes a 96-well plate coated with capture antibodies detection antibodies calibrators controls 10X read buffer 10X wash buffer and plate seals The assay takes approximately 3 5 hours to complete
Catalog No. 50-000-78594 Supplier Meso Scale Discovery Supplier No. K1535VK24
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX Rat GLP-1 (Inactive) Assay Kit is a multiplex immunoassay that measures GLP-1 (7-36) amide and GLP-1 (9-36) amide in rat serum plasma and cell culture supernatants The assay uses electrochemiluminescence detection technology and provides high sensitivity broad dynamic range and rapid results This assay allows for the simultaneous detection of both active and inactive forms of GLP-1 enabling more complete characterization of GLP-1 biology The kit includes a 96-well plate coated with capture antibodies detection antibodies calibrators controls 10X read buffer 10X wash buffer and plate seals The assay takes approximately 3 5 hours to complete