Learn More
Meso Scale Discovery UPLEX Rat GLP1 inactive ASYQP
SDP
Catalog No. 5000082189
Encompass
The Meso Scale Discovery U-PLEX Rat GLP-1 (Inactive) Assay Kit is a multiplex immunoassay that measures GLP-1 (7-36) amide and GLP-1 (9-36) amide in rat serum plasma and cell culture supernatants The assay uses electrochemiluminescence detection technology and provides high sensitivity broad dynamic range and rapid results This quantitative assay can be used to measure GLP-1 levels and assess GLP-1 biology and metabolism The kit includes a 96-well plate coated with capture antibodies detection antibodies calibrators controls 10X diluent read buffer and wash buffer
Catalog No. 50-000-82189 Supplier Meso Scale Discovery Supplier No. K1535VK22
The Meso Scale Discovery U-PLEX Rat GLP-1 (Inactive) Assay Kit is a multiplex immunoassay that measures GLP-1 (7-36) amide and GLP-1 (9-36) amide in rat serum plasma and cell culture supernatants The assay uses electrochemiluminescence detection technology and provides high sensitivity broad dynamic range and rapid results This quantitative assay can be used to measure GLP-1 levels and assess GLP-1 biology and metabolism The kit includes a 96-well plate coated with capture antibodies detection antibodies calibrators controls 10X diluent read buffer and wash buffer