Learn More
Meso Scale Discovery UPLEX Rat GLP1 TTL ASY
SDP
Catalog No. 5000082139
Encompass
The Meso Scale Discovery U-PLEX Rat GLP-1 Total Assay Kit is an electrochemiluminescent immunoassay for the quantitative measurement of total GLP-1 in rat serum plasma and cell culture supernatants The assay uses MSD GOLD plate technology and provides high sensitivity a broad dynamic range and requires a low sample volume of 25 L The lower limit of detection is 1 15 pg/mL The assay measures active GLP-1 as well as the primary metabolite GLP-1 (9-36) amide It is for research use only
Catalog No. 50-000-82139 Supplier Meso Scale Discovery Supplier No. K1535UK4
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX Rat GLP-1 Total Assay Kit is an electrochemiluminescent immunoassay for the quantitative measurement of total GLP-1 in rat serum plasma and cell culture supernatants The assay uses MSD GOLD plate technology and provides high sensitivity a broad dynamic range and requires a low sample volume of 25 L The lower limit of detection is 1 15 pg/mL The assay measures active GLP-1 as well as the primary metabolite GLP-1 (9-36) amide It is for research use only