Learn More
Meso Scale Discovery UPLEX Rat GLP1 TTL ASY384well
SDP
Catalog No. 5000081370
Encompass
The Meso Scale Discovery U-PLEX Rat GLP-1 Total 384-well Assay is an electrochemiluminescent immunoassay designed to measure total GLP-1 levels in rat serum plasma and cell culture supernatants This 384-well multiplex assay utilizes MSD s proprietary SULFO-TAG labels and multi-array technology to provide sensitive and reproducible results with a broad dynamic range and low sample volume requirements The assay can detect GLP-1 levels between 0 06 - 1000 pg/mL and includes reagents for 384 tests This quantitative immunoassay provides a streamlined workflow and rapid results to monitor total GLP-1 levels within a single sample
Catalog No. 50-000-81370 Supplier Meso Scale Discovery Supplier No. K2535UK2
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX Rat GLP-1 Total 384-well Assay is an electrochemiluminescent immunoassay designed to measure total GLP-1 levels in rat serum plasma and cell culture supernatants This 384-well multiplex assay utilizes MSD s proprietary SULFO-TAG labels and multi-array technology to provide sensitive and reproducible results with a broad dynamic range and low sample volume requirements The assay can detect GLP-1 levels between 0 06 - 1000 pg/mL and includes reagents for 384 tests This quantitative immunoassay provides a streamlined workflow and rapid results to monitor total GLP-1 levels within a single sample