Learn More
Meso Scale Discovery UPLEX Rat GLP1 TTL ASY384well
SDP
Catalog No. 5000081371
Encompass
The Meso Scale Discovery U-PLEX Rat GLP-1 Total 384-well Assay is an electrochemiluminescent immunoassay for measuring total GLP-1 in rat serum plasma and other samples This 384-well assay uses MSD s SULFO-TAG labels and multi-array technology for detecting multiple protein biomarkers in one sample It has a detection range of 1 56 - 400 pg/mL and includes a plate with capture antibodies detection antibodies with ruthenium read buffer wash buffer blocker solutions calibrators and controls The assay offers high sensitivity low sample volume and short assay times
Catalog No. 50-000-81371 Supplier Meso Scale Discovery Supplier No. K2535UK4
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX Rat GLP-1 Total 384-well Assay is an electrochemiluminescent immunoassay for measuring total GLP-1 in rat serum plasma and other samples This 384-well assay uses MSD s SULFO-TAG labels and multi-array technology for detecting multiple protein biomarkers in one sample It has a detection range of 1 56 - 400 pg/mL and includes a plate with capture antibodies detection antibodies with ruthenium read buffer wash buffer blocker solutions calibrators and controls The assay offers high sensitivity low sample volume and short assay times