Learn More
Meso Scale Discovery UPLEX Rat GLP1 TTL ASYQP
SDP
Catalog No. 5000080354
Encompass
The Meso Scale Discovery U-PLEX Rat GLP-1 Total Assay Kit is an electrochemiluminescent immunoassay for the quantitative measurement of total GLP-1 in rat serum plasma and cell culture supernatants The assay uses MSD GOLD plate technology and provides high sensitivity wide dynamic range and low sample volume requirements This multiplex kit includes a plate coated with capture antibodies detection antibodies labeled with MSD SULFO-TAG calibrator and sample diluent The kit can accurately quantify GLP-1 levels between 0 61 pg/mL to 2500 pg/mL with an assay time of 3 5 hours
Catalog No. 50-000-80354 Supplier Meso Scale Discovery Supplier No. K1535UK24
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX Rat GLP-1 Total Assay Kit is an electrochemiluminescent immunoassay for the quantitative measurement of total GLP-1 in rat serum plasma and cell culture supernatants The assay uses MSD GOLD plate technology and provides high sensitivity wide dynamic range and low sample volume requirements This multiplex kit includes a plate coated with capture antibodies detection antibodies labeled with MSD SULFO-TAG calibrator and sample diluent The kit can accurately quantify GLP-1 levels between 0 61 pg/mL to 2500 pg/mL with an assay time of 3 5 hours