Learn More
Meso Scale Discovery UPLEX Rat GLPactive ASY384
SDP
Catalog No. 5000079468
Encompass
The Meso Scale Discovery U-PLEX Rat GLP-1 (Active) Assay is a 384-well electrochemiluminescent immunoassay for measuring active GLP-1 in rat serum plasma and cell culture supernatants It uses MSD s SULFO-TAG labels and multi-array technology for sensitive detection The kit includes a 384-well plate with a GLP-1 capture antibody detection antibody with an electrochemiluminescent label calibrator solutions and necessary buffers and washes Active GLP-1 is quantified in 3 5 hours using small samples enabling high-throughput research analysis
Catalog No. 50-000-79468 Supplier Meso Scale Discovery Supplier No. K2536LK4
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX Rat GLP-1 (Active) Assay is a 384-well electrochemiluminescent immunoassay for measuring active GLP-1 in rat serum plasma and cell culture supernatants It uses MSD s SULFO-TAG labels and multi-array technology for sensitive detection The kit includes a 384-well plate with a GLP-1 capture antibody detection antibody with an electrochemiluminescent label calibrator solutions and necessary buffers and washes Active GLP-1 is quantified in 3 5 hours using small samples enabling high-throughput research analysis