Learn More
Meso Scale Discovery UPLEX Rat Glucagon ASY 25 PL
SDP
Catalog No. 5000078591
Encompass
The Meso Scale Discovery U-PLEX Rat Glucagon Assay Kit is an electrochemiluminescent immunoassay for the quantitative measurement of glucagon in rat serum plasma and cell culture supernatants The assay uses MSD s proprietary SULFO-TAG labels and multi-array technology to provide sensitive and reproducible results with low sample volume requirements The kit includes a 96-well plate coated with capture antibody detection antibody conjugated to ruthenium label calibrators and sample diluent The assay has a typical sensitivity of 0 3 pg/mL and an upper limit of quantification of 1000 pg/mL
Catalog No. 50-000-78591 Supplier Meso Scale Discovery Supplier No. K1535YK4
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX Rat Glucagon Assay Kit is an electrochemiluminescent immunoassay for the quantitative measurement of glucagon in rat serum plasma and cell culture supernatants The assay uses MSD s proprietary SULFO-TAG labels and multi-array technology to provide sensitive and reproducible results with low sample volume requirements The kit includes a 96-well plate coated with capture antibody detection antibody conjugated to ruthenium label calibrators and sample diluent The assay has a typical sensitivity of 0 3 pg/mL and an upper limit of quantification of 1000 pg/mL