Learn More
Meso Scale Discovery UPLEX Rat Glucagon ASY384well
SDP
Catalog No. 5000079344
Encompass
The Meso Scale Discovery U-PLEX Rat Glucagon 384-Well Assay Kit is a multiplex immunoassay that measures glucagon levels in rat serum plasma and cell culture supernatants The kit uses electrochemiluminescence detection technology and provides a 384-well plate formatted assay with high sensitivity and broad dynamic range The assay requires a low sample volume of 25 L and has a testing time of approximately 3 5 hours The lower limit of detection is 1 1 pg/mL This assay kit offers a streamlined workflow and is suitable for measuring glucagon levels in rat samples for research applications
Catalog No. 50-000-79344 Supplier Meso Scale Discovery Supplier No. K2535YK2
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX Rat Glucagon 384-Well Assay Kit is a multiplex immunoassay that measures glucagon levels in rat serum plasma and cell culture supernatants The kit uses electrochemiluminescence detection technology and provides a 384-well plate formatted assay with high sensitivity and broad dynamic range The assay requires a low sample volume of 25 L and has a testing time of approximately 3 5 hours The lower limit of detection is 1 1 pg/mL This assay kit offers a streamlined workflow and is suitable for measuring glucagon levels in rat samples for research applications