Learn More
Meso Scale Discovery UPLEX Rat Glucagon ASY384well
SDP
Catalog No. 5000081156
Encompass
The Meso Scale Discovery U-PLEX Glucagon Assay Kit is an electrochemiluminescent immunoassay for measuring glucagon in human serum plasma and cell culture supernatants It uses MSD s SULFO-TAG labels and multi-array plate technology for sensitive detection The kit includes a 96-well plate with capture antibody detection antibody with ruthenium label calibrators and diluent Sensitivity is 0 24 pg/mL with a quantification limit of 1000 pg/mL The assay takes about 3 5 hours allowing high-throughput glucagon quantification from various samples
Catalog No. 50-000-81156 Supplier Meso Scale Discovery Supplier No. K2535YK4
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX Glucagon Assay Kit is an electrochemiluminescent immunoassay for measuring glucagon in human serum plasma and cell culture supernatants It uses MSD s SULFO-TAG labels and multi-array plate technology for sensitive detection The kit includes a 96-well plate with capture antibody detection antibody with ruthenium label calibrators and diluent Sensitivity is 0 24 pg/mL with a quantification limit of 1000 pg/mL The assay takes about 3 5 hours allowing high-throughput glucagon quantification from various samples