Learn More
Meso Scale Discovery UPLEX Rat Glucagon ASYQP
SDP
Catalog No. 5000081891
Encompass
The Meso Scale Discovery U-PLEX Rat Glucagon Assay Kit is an electrochemiluminescent immunoassay for the quantitative measurement of glucagon in rat serum plasma and cell culture supernatants The assay uses MSD s proprietary SULFO-TAG labels and multi-array technology to provide sensitive and rapid detection of glucagon on the ultra-sensitive MESO QuickPlex SQ 120 instrument The assay has a detection range of 1 56 - 400 pg/mL and exhibits no cross-reactivity with other hormones The kit includes a 96-well plate coated with capture antibody detection antibody conjugated to ruthenium label calibrators and sample diluent The total assay time is 3 5 hours
Catalog No. 50-000-81891 Supplier Meso Scale Discovery Supplier No. K1535YK24
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX Rat Glucagon Assay Kit is an electrochemiluminescent immunoassay for the quantitative measurement of glucagon in rat serum plasma and cell culture supernatants The assay uses MSD s proprietary SULFO-TAG labels and multi-array technology to provide sensitive and rapid detection of glucagon on the ultra-sensitive MESO QuickPlex SQ 120 instrument The assay has a detection range of 1 56 - 400 pg/mL and exhibits no cross-reactivity with other hormones The kit includes a 96-well plate coated with capture antibody detection antibody conjugated to ruthenium label calibrators and sample diluent The total assay time is 3 5 hours