Learn More
Meso Scale Discovery UPLEX Rat Leptin ASY384well
SDP
Catalog No. 5000080010
Encompass
The Meso Scale Discovery U-PLEX Rat Leptin 384-Well Assay Kit is a multiplex immunoassay that measures leptin in rat serum plasma and cell culture supernatants The kit uses electrochemiluminescence detection technology and provides a 384-well plate formatted assay allowing high throughput analysis of leptin The assay has high sensitivity wide dynamic range and requires low sample volume This product allows for the quantitative measurement of leptin which plays a key role in regulating energy intake and expenditure
Catalog No. 50-000-80010 Supplier Meso Scale Discovery Supplier No. K2535ZK4
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX Rat Leptin 384-Well Assay Kit is a multiplex immunoassay that measures leptin in rat serum plasma and cell culture supernatants The kit uses electrochemiluminescence detection technology and provides a 384-well plate formatted assay allowing high throughput analysis of leptin The assay has high sensitivity wide dynamic range and requires low sample volume This product allows for the quantitative measurement of leptin which plays a key role in regulating energy intake and expenditure