Learn More
Meso Scale Discovery UPLEX Rat PYY TTL ASY
SDP
Catalog No. 5000082184
Encompass
The Meso Scale Discovery U-PLEX Rat PYY Total Assay Kit is a multiplex immunoassay for the quantitative measurement of total PYY in rat serum plasma and cell culture supernatants The assay uses electrochemiluminescence (ECL) detection technology and provides high sensitivity broad dynamic range and intra- and inter-assay precision This kit includes a 96-well plate coated with capture antibodies detection antibodies labeled with MSD SULFO-TAG calibrator and sample diluent The assay has a typical sensitivity of 0 3 pg/mL and an upper limit of quantification of 10 000 pg/mL
Catalog No. 50-000-82184 Supplier Meso Scale Discovery Supplier No. K1536BK4
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX Rat PYY Total Assay Kit is a multiplex immunoassay for the quantitative measurement of total PYY in rat serum plasma and cell culture supernatants The assay uses electrochemiluminescence (ECL) detection technology and provides high sensitivity broad dynamic range and intra- and inter-assay precision This kit includes a 96-well plate coated with capture antibodies detection antibodies labeled with MSD SULFO-TAG calibrator and sample diluent The assay has a typical sensitivity of 0 3 pg/mL and an upper limit of quantification of 10 000 pg/mL